Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1-Penten-3-ol

Catalog No. P86025G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P86025G 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P86025G Supplier Sigma Aldrich Supplier No. P86025G
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 3-methoxy-1-butanol can be used as:
  • A solvent in the synthesis of potassium 3-methoxy-1-butylxanthate, an intermediate used in the preparation of xanthate complexes
  • A model compound in the study of dehydration of alcohols using ceo2
  • As a catalyst
  • An intermediate to synthesize perylene diimide-based dyes applicable as the colorant of the black matrix

Specifications

CAS 616-25-1
Density 0.838 g/mL at 20°C (literature); 0.839 g/mL (at 25°C (literature))
Boiling Point 114°C to 115°C (lit.)
Molecular Formula C5H10O
Refractive Index n20/D 1.424 (literature)
Linear Formula CH3CH2CH(OH)CH=CH2
MDL Number MFCD00004573
Quantity 1 L
Synonym Ethyl vinyl carbinol
Molecular Weight (g/mol) 86.13
Percent Purity 99%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.