Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Penten-3-ol 97.0+%, TCI America™
SDP

Catalog No. P104325ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P104325ML 25 mL
P10435ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P104325ML Supplier TCI America Supplier No. P104325ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 616-25-1
Molecular Formula C5H10O
Molecular Weight (g/mol) 86.13
MDL Number MFCD00004573
InChI Key VHVMXWZXFBOANQ-UHFFFAOYNA-N
Synonym 1-penten-3-ol, ethyl vinyl carbinol, vinyl ethyl carbinol, 1-pentene-3-ol, 1-ethylallyl alcohol, alpha-ethylallyl alcohol, 1-penten-3-ol e, 1-penten-3-ol, r, fema no. 3584, ethylvinylcarbinol
PubChem CID 12020
IUPAC Name pent-1-en-3-ol
SMILES CCC(O)C=C

Specifications

Color Colorless
Boiling Point 37°C
Quantity 25 mL
UN Number 1987
Formula Weight 86.13
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Penten-3-ol
EINECSNumber (2)-2393
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.