Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1-Pentene

Catalog No. 7697125ML Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
25 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
76-971-25ML 25 mL
76-971-100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 76-971-25ML Supplier Sigma Aldrich Supplier No. 7697125ML
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 1-pentene can be used as a component in gasoline surrogates
  • It can also be used in the synthesis of (+/-)-byssochlamic acid and rugulactone

Specifications

CAS 109-67-1
Density 0.641 g/mL (at 25°C (literature))
Boiling Point 29.9°C to 30.1°C (lit.)
Molecular Formula C5H10
Refractive Index n20/D 1.371 (literature); n20/D 1.371
Linear Formula CH3CH2CH2CH=CH2
MDL Number MFCD00003567
Quantity 25 mL
Molecular Weight (g/mol) 70.13
Percent Purity ≥98.5% (GC)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.