Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Pentene, ≥99.5% (GC), MilliporeSigma™ Supelco™

Catalog No. 111018955 Shop All MilliporeSigma Supelco Products
Change view
Click to view available options
Quantity:
5 mL
50 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11-101-8955 5 mL
11-101-8528 50 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 11-101-8955 Supplier MilliporeSigma Supelco Supplier No. 769695ML
Add to Cart
Add to Cart
Featured Industries: Cleaning Products, Cosmetics, Food and Beverages, Personal Care and Petroleum
TRUSTED_SUSTAINABILITY

Specifications

Ignition Point 522°F
CAS 109-67-1
Vapor Pressure 10.27 psi (20°C)
Vapor Density 2.4 (vs. air)
Density 0.641 g/mL (at 25°C)
Flash Point -51°C - Closed Cup
Physical Form Neat
Linear Formula CH3 CH2 CH2 CH=CH2
MDL Number MFCD00003567
Boiling Point 29.9°C to 30.1°C
Type Analytical Standard
Quantity 5 mL
Formula Weight 70.13
Percent Purity ≥99.5% (GC)
Grade Analytical Standard
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.