Learn More
Ready America™ 1 Person 3 Day Elite Emergency Backpack
SDP
Catalog No. 17467153
Change view
Click to view available options
Color:
Black
Catalog No. Color
17-467-153 Black
1 options
Catalog No. 17-467-153 Supplier Ready America™ Supplier No. 70450
Add to Cart
Add to Cart
This 1-Person Elite Emergency Kit and backpack is perfect for the individual that wants to be prepared and ready for almost anything. It is recommended that we have at least 72-hours of emergency food and water, and this 3-day backpack has that and more
  • First responders advise us that we should be prepared for at least the first 72 hours after a disaster, it is absolutely essential that we have emergency supplies at our disposal. That is why Ready America has developed the Elite 1 Person Emergency backpack with the essentials you need for 3 days. It contains all the life saving supplies and more that you will need to survive for those crucial 72 hours.
  • The Emergency Power Station is a four-function hand-crank unit that includes a flashlight, AM/FM radio, cell phone charger and a siren. One minute of cranking produces either 30 minutes of bright light, 2 minutes of talk time on a cell phone, or 15 minutes of radio. The Heavy Duty Multi-Tool includes: knife, screwdriver, bottle opener, saw, fire starter and pliers
  • Emergency Kit Contains:
    • (1) Food Bar (3600 Calorie) (5-year shelf life)
    • (9) Pouches of Water (4.225 oz.) (5-year shelf life)
    • (1) Potable Aqua Germicidal Tablets (50 count)
    • (3) Bio Hazard Bags
    • (2) 10 in. Nitrile Gloves
    • (1) Heavy Duty Work Gloves
    • (1) Niosh N-95 Approved Dust Mask
    • (1) First-Aid Kit (33 piece)
    • (1) Heavy Duty Multi-Tool
    • (1) 8 in. Survival Knife with Compass & Survival Kit (matches, needle/thread, fishing line/hook)
    • (1) Duct Tape (10 yards)
    • (1) 5 in 1 Survival Whistle
    • (1) Emergency Poncho
    • (1) Emergency Survival Blanket
    • (1) Knit Cap (Beanie)
    • (1) Pocket Tissue
    • (3) Wet Naps
    • (1) Wipes in Soft Dispenser
    • (1) Emergency Power Station 4 Function
    • (1) 12 Hour Light Stick
    • (1) Water Bottle (1 Liter)
    • (1) Hygiene Kit Basic
    • (1) 25 in. Cord
    • (1) Document Pouch
    • (1) Large Elite Backpack
Specifications
Product Type Emergency Backpack
Color Black
Disposable Reusable