Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Phenethylimidazole 98.0+%, TCI America™
SDP

Catalog No. P20655G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P20655G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P20655G Supplier TCI America Supplier No. P20655G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 49823-14-5
Molecular Formula C11H12N2
Molecular Weight (g/mol) 172.231
MDL Number MFCD11977663
InChI Key SSENHTOBLYRWKU-UHFFFAOYSA-N
Synonym 1-(Phenylethyl)imidazole
PubChem CID 10219621
IUPAC Name 1-(2-phenylethyl)imidazole
SMILES C1=CC=C(C=C1)CCN2C=CN=C2

Specifications

Melting Point 34°C
Color White-Yellow
Boiling Point 145°C
Quantity 5 g
Formula Weight 172.23
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Lumps
Chemical Name or Material 1-Phenethylimidazole
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.