Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1% Phenolphthalein in 50% Isopropanol Indicator 5, Reagents

Catalog No. 01334933
Change view
Click to view available options
Quantity:
500 mL
1 L
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-334-933 500 mL
01-334-934 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-334-933 Supplier Reagents Holdings Llc Supplier No. CI161800500A
Add to Cart
Add to Cart

1% Phenolphthalein in 50% Isopropanol Indicator 5, Reagent, for general laboratory use. Manufactured in ISO 9001 facility.

Specifications

Chemical Name or Material 1% Phenolphthalein in 50% Isopropanol Indicator 5
Boiling Point 82°C
CAS 7732-18-5, 67-63-0
Molecular Formula C20H14O4/C3H7OH/H2O
Synonym 3,3-Bis(4-hydroxyphenyl)isobenzofuran-1(3H)-one
Percent Purity 1%
Grade Reagent
Packaging Poly Bottle
Specific Gravity 0.9
Color Colorless
pH <8
Quantity 500 mL
Physical Form Liquid
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.