Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Phenoxy-2-chloropropane 97.0+%, TCI America™
SDP

Catalog No. P011710G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P011710G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P011710G Supplier TCI America Supplier No. P011710G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 53491-30-8
Molecular Formula C9H11ClO
Molecular Weight (g/mol) 170.636
MDL Number MFCD00059488
InChI Key LTRAIWUUCBYXHZ-UHFFFAOYSA-N
Synonym 2-Chloropropyl Phenyl Ether
PubChem CID 565152
IUPAC Name 2-chloropropoxybenzene
SMILES CC(COC1=CC=CC=C1)Cl

Specifications

Color Yellow
Boiling Point 95°C
Quantity 10 g
Formula Weight 170.64
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Phenoxy-2-chloropropane
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.