Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1-Phenoxy-2-Propanol

Catalog No. 4844231L Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
48-442-31L 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Warning
Catalog No. 48-442-31L Supplier Sigma Aldrich Supplier No. 4844231L
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

1-Phenoxy-2-propanol (PP), a gycol ether, can be synthesized by reacting propylene oxide with phenol in the presence of Al2O3-MgO/Fe3O4 catalyst. The influence of its anesthetic property on gastropods has been analyzed. Its degradation by microorganisms in different soil types has been investigated.1-Phenoxy-2-propanol has been used as an additive to saline to induce anesthetic effect on the peripheral tissues isolated from Biomphalaria alexandrina prior to fixation.DOWANOL is a registered trademark of The Dow Chemical Company or an affiliated company of Dow

Specifications

CAS 98437-24-2
Melting Point 77°C to 83°C
Molecular Formula C19H17P
Refractive Index n20/D 1.523
Linear Formula C6H5CH2P(C6H5)2
MDL Number MFCD00014083
Quantity 5 g
Synonym DOWANOL(R) PPh; Propylene glycol phenyl ether; Propylene phenoxytol
Molecular Weight (g/mol) 276.31 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.