Learn More
Medchemexpress LLC 1-phenyl-1,2-ethanediol | 93-56-1 | MFCD00003546 | 99.7% | 138.17 g/mol | C8H10O2 | 1 ML
SDP
Catalog No. 5000300019
Encompass_Preferred
1-Phenyl-1,2-ethanediol (styrene glycol) is a benzyl diol and a primary metabolite of styrene. The material is supplied as a white to light brown solid or as a ready-to-use 10 mM solution in DMSO (1 mL). Certificate of analysis reports high purity and provides batch-specific analytical data for quality assurance.
  • High purity: 99.74% (HPLC).
  • Supplied as a 10 mM solution in DMSO (1 mL) for convenience.
  • Also available as solid in multiple pack sizes for bulk use.
  • Suitable for analytical standards, reference, and metabolic studies.
  • Certificate of analysis available for batch-specific verification.
Catalog No. 50-003-00019 Supplier Medchemexpress LLC Supplier No. HYW01578810MM/1ML
1-Phenyl-1,2-ethanediol (styrene glycol) is a benzyl diol and a primary metabolite of styrene. The material is supplied as a white to light brown solid or as a ready-to-use 10 mM solution in DMSO (1 mL). Certificate of analysis reports high purity and provides batch-specific analytical data for quality assurance.
  • High purity: 99.74% (HPLC).
  • Supplied as a 10 mM solution in DMSO (1 mL) for convenience.
  • Also available as solid in multiple pack sizes for bulk use.
  • Suitable for analytical standards, reference, and metabolic studies.
  • Certificate of analysis available for batch-specific verification.