Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Phenyl-1,2-ethanediol, 97%

Catalog No. p-3779419
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Packaging:
Glass bottle
Plastic bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC130590050 5 g Glass bottle
AC130590250 25 g Plastic bottle
AC130591000 100 g Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC130590050 Supplier Thermo Scientific Chemicals Supplier No. 130590050
Only null left
Add to Cart

CAS: 93-56-1 | C8H10O2 | 138.17 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 93-56-1
Molecular Formula C8H10O2
Molecular Weight (g/mol) 138.17
MDL Number MFCD00003546
InChI Key PWMWNFMRSKOCEY-UHFFFAOYSA-N
Synonym 1-phenyl-1,2-ethanediol, styrene glycol, phenylethylene glycol, phenylethanediol, phenyl glycol, 1,2-ethanediol, 1-phenyl, phenyl-1,2-ethanediol, styrolyl alcohol, fenylglycol, 1-phenylethylene glycol
PubChem CID 7149
IUPAC Name 1-phenylethane-1,2-diol
SMILES C1=CC=C(C=C1)C(CO)O

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 61°C to 67°C
Color White to Beige
Boiling Point 272°C to 274°C
Infrared Spectrum Authentic
Assay Percent Range 96% min. (GC)
Packaging Glass bottle
Linear Formula HOCH2CH(C6H5)OH
Quantity 5 g
Beilstein 06, 907
Merck Index 15, 8991
Formula Weight 138.17
Percent Purity 97%
Physical Form Crystalline Powder or Flakes
Chemical Name or Material 1-Phenyl-1, 2-ethanediol
Show More Show Less
EINECSNumber 202-258-1

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.