Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1-phenyl-1,2-propanedione | 579-07-7 | MFCD00008755 | 99.8% | 148.16 g/mol | C9H8O2 | 50 G
SDP

Catalog No. 5000301303
Encompass_Preferred

1-Phenyl-1,2-propanedione is an alpha-diketone research compound reported as a biosynthetic precursor of ephedrine alkaloids. It is supplied as a characterized analytical standard for laboratory use, with documented physicochemical properties and recommended storage conditions for stability.

  • High purity suitable for analytical and research applications.
  • Molecular formula C9H8O2; molecular weight 148.16 g/mol.
  • Soluble in DMSO at ≥200 mg/mL; ultrasonic assistance may be required.
  • Light yellow to yellow liquid; density ~1.101 g/cm3.
  • Store protected from light and under inert gas; cold storage recommended for solutions.

Catalog No. 50-003-01303 Supplier Medchemexpress LLC Supplier No. HYW01875850G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

1-Phenyl-1,2-propanedione is an alpha-diketone research compound reported as a biosynthetic precursor of ephedrine alkaloids. It is supplied as a characterized analytical standard for laboratory use, with documented physicochemical properties and recommended storage conditions for stability.

  • High purity suitable for analytical and research applications.
  • Molecular formula C9H8O2; molecular weight 148.16 g/mol.
  • Soluble in DMSO at ≥200 mg/mL; ultrasonic assistance may be required.
  • Light yellow to yellow liquid; density ~1.101 g/cm3.
  • Store protected from light and under inert gas; cold storage recommended for solutions.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.