Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1-phenyl-1-butyne-d5 | 1219803-38-9 | >98.0% | 135.22 g/mol | C10H5D5 | 5 MG
SDP

Catalog No. 5000283027
Encompass_Preferred

1-Butynylbenzene-d5 is the deuterium-labeled analog of 1-butynylbenzene (1-phenyl-1-butyne). It is supplied as a small-quantity, isotopically labeled research reagent containing an alkyne functional group suitable for copper-catalyzed azide-alkyne cycloaddition (CuAAC) and for use as an internal standard in analytical applications.

  • Deuterium-labeled alkyne reagent.
  • CAS number: 1219803-38-9.
  • Molecular formula C10H5D5; molecular weight 135.22 g/mol.
  • Purity >98.0% chemical purity.
  • Appearance liquid.
  • Pack size 5 MG.
  • Storage pure form -20°C; in solvent -80°C (6 months) or -20°C (1 month).

Catalog No. 50-002-83027 Supplier Medchemexpress LLC Supplier No. HYW142175S5MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

1-Butynylbenzene-d5 is the deuterium-labeled analog of 1-butynylbenzene (1-phenyl-1-butyne). It is supplied as a small-quantity, isotopically labeled research reagent containing an alkyne functional group suitable for copper-catalyzed azide-alkyne cycloaddition (CuAAC) and for use as an internal standard in analytical applications.

  • Deuterium-labeled alkyne reagent.
  • CAS number: 1219803-38-9.
  • Molecular formula C10H5D5; molecular weight 135.22 g/mol.
  • Purity >98.0% chemical purity.
  • Appearance liquid.
  • Pack size 5 MG.
  • Storage pure form -20°C; in solvent -80°C (6 months) or -20°C (1 month).

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.