Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Phenyl-1-cyclohexene 95.0+%, TCI America™
SDP

Catalog No. P130225ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 mL
25 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P130225ML 25 mL
P13025ML 5 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P130225ML Supplier TCI America Supplier No. P130225ML
Add to Cart
Add to Cart

Chemical Identifiers

CAS 771-98-2
Molecular Formula C12H14
Molecular Weight (g/mol) 158.244
MDL Number MFCD00001542
InChI Key WCMSFBRREKZZFL-UHFFFAOYSA-N
Synonym 1-phenyl-1-cyclohexene, 1-phenylcyclohexene, 1-phenylcyclohex-1-ene, benzene, 1-cyclohexen-1-yl, phenylcyclohexene, cyclohexenylbenzene, benzene, cyclohexenyl, 2,3,4,5-tetrahydro-1,1'-biphenyl, cyclohexene, 1-phenyl, trans-1-phenylcyclohexene
PubChem CID 13043
IUPAC Name cyclohexen-1-ylbenzene
SMILES C1CCC(=CC1)C2=CC=CC=C2

Specifications

Color Colorless
Boiling Point 252°C
Quantity 25 mL
Formula Weight 158.24
Percent Purity ≥95.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Phenyl-1-cyclohexene
RTECSNumber GW6600000
TSCA No
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.