Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Phenyl-1-pentyne 97.0+%, TCI America™
SDP

Catalog No. P035925ML Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P035925ML 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. P035925ML Supplier TCI America Supplier No. P035925ML

missing translation for 'validMainframeMsg'

Specifications

CAS 4250-81-1
Color Yellow
Boiling Point 214°C
Molecular Formula C11H12
MDL Number MFCD00039959
Quantity 25 mL
Synonym 1-phenyl-1-pentyne, pent-1-yn-1-ylbenzene, 1-pentynylbenzene, benzene, 1-pentynyl, 1-pentynylbenzene #, acmc-1aso7, 1-phenyl-2-propylacetylene
InChI Key DEGIOKWPYFOHGH-UHFFFAOYSA-N
SMILES CCCC#CC1=CC=CC=C1
IUPAC Name (pent-1-yn-1-yl)benzene
Molecular Weight (g/mol) 144.22
PubChem CID 77923
Formula Weight 144.22
Percent Purity ≥97.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Phenyl-1-pentyne
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.