Learn More
Medchemexpress LLC 1-Phenyl-1-propanol | 93-54-9 | MFCD00004564 | 99.9% | 136.19 | 50 G
SDP
Catalog No. 5000453880
Encompass_Preferred
1-Phenyl-1-propanol serves as a versatile chiral building block and synthetic intermediate within the pharmaceutical industry. It is specifically utilized in the synthesis of the anti-depressant agent Fluoxetine.
  • Chiral building block
  • Synthetic intermediate for pharmaceuticals
  • Intermediate for anti-depressant agent Fluoxetine
Catalog No. 50-004-53880 Supplier Medchemexpress LLC Supplier No. HYW01261850G
1-Phenyl-1-propanol serves as a versatile chiral building block and synthetic intermediate within the pharmaceutical industry. It is specifically utilized in the synthesis of the anti-depressant agent Fluoxetine.
  • Chiral building block
  • Synthetic intermediate for pharmaceuticals
  • Intermediate for anti-depressant agent Fluoxetine