Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(+/-)-1-Phenyl-1-propanol, 98+%

Catalog No. p-7081033
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1083014 25 g
AAA1083022 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1083014 Supplier Thermo Scientific Chemicals Supplier No. A1083014
Only null left
Add to Cart

CAS: 93-54-9 | C9H12O | 136.194 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 93-54-9
Molecular Formula C9H12O
Molecular Weight (g/mol) 136.194
MDL Number MFCD00004564
InChI Key DYUQAZSOFZSPHD-UHFFFAOYSA-N
Synonym 1-phenyl-1-propanol, phenycholon, fepar, ejibil, felicur, livonal, 1-phenylpropanol, gallenperlen, phenylchol, bilergon
PubChem CID 7147
IUPAC Name 1-phenylpropan-1-ol
SMILES CCC(C1=CC=CC=C1)O

Specifications

Density 0.994
Boiling Point 217°C to 219°C
Flash Point 95°C (203°F)
Refractive Index 1.52
Quantity 25 g
Beilstein 1906759
Merck Index 14,3768
Formula Weight 136.19
Percent Purity ≥98%
Chemical Name or Material (±)-1-Phenyl-1-propanol
Hazard Category H227-H302
Hazard Statement GHS H Statement
H302
Harmful if swallowed.
Precautionary Statement P210-P264b-P270-P280-P301+P312-P330-P370+P378q-P501c
EINECSNumber 202-256-0
RTECSNumber DO5470000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.