Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1-Phenyl-1-propyne

Catalog No. 16124125G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
16-124-125G 25 g
16-124-15G 5 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 16-124-125G Supplier Sigma Aldrich Supplier No. 16124125G
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Co-catalyzed reaction between cyclopentene and 1-phenyl-1-propyne has been reported
  • Bonding properties of 1-phenyl-1-propyne on cu(111) at 100k have been studied using temperature-programmed desorption and x-ray, ultraviolet and two-photon photoemission spectroscopies
  • 1-phenyl-1-propyne is an inhibitor of dopamine beta-hydroxylase
  • Polymerization of 1-phenyl-1-propyne by tacl5 and nbcl5 has been reported

Specifications

CAS 673-32-5
Density 0.928 g/mL (at 25°C (literature))
Boiling Point 185°C (lit.)
Molecular Formula C9H8
Refractive Index n20/D 1.564 (literature)
Linear Formula C6H5C///CCH3
MDL Number MFCD00009272
Quantity 25 g
Molecular Weight (g/mol) 116.16
Percent Purity 99%
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.