Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1-Phenyl-1-trimethylsiloxyethylene

Catalog No. 2350405G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
23-504-05G 5 g
23-504-025G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 23-504-05G Supplier Sigma Aldrich Supplier No. 2350405G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 1-phenyl-1-trimethylsiloxyethylene is a styrene type silyl enol ether, reacts with formaldehyde and 2,4-pentanedione to yield the corresponding dihydropyran. it also undergoes mukaiyama aldol reaction with aldehydes in water in the presence of amphiphilic calix[6]arene derivatives as surfactants.1-phenyl-1-trimethylsiloxyethylene has been used in the synthesis of:
  • α-amino ketones in water via mannich-type reaction
  • Cis-2,6-disubstituted dihydropyrans via three-component, one-pot cascade reaction

Specifications

CAS 13735-81-4
Density 0.938 g/mL (at 25°C (literature))
Boiling Point 88°C to 89°C (11 mmHg)
Molecular Formula C11H16OSi
Refractive Index n20/D 1.502 (literature)
Linear Formula C6H5C(=CH2)OSi(CH3)3
MDL Number MFCD00008582
Quantity 5 g
Synonym alpha-(Trimethylsiloxy)styrene; Acetophenone enol trimethylsilyl ether
Molecular Weight (g/mol) 192.33
Percent Purity 98%
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.