Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-phenyl-1H-imidazole-2-carbaldehyde, Thermo Scientific™

Catalog No. CC75904DE
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
Packaging:
Amber Glass Bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC75904DE 5 g Amber Glass Bottle
CC75904CB 250 mg Amber Glass Bottle
CC75904DA 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. CC75904DE Supplier Thermo Scientific Chemicals Supplier No. CC75904DE
Add to Cart

missing translation for 'validMainframeMsg'

Chemical Identifiers

CAS 6002-15-9
Molecular Formula C10H8N2O
Molecular Weight (g/mol) 172.187
InChI Key VJDRBJPZIHUDNE-UHFFFAOYSA-N
Synonym 1-phenyl-1h-imidazole-2-carbaldehyde, 1h-imidazole-2-carboxaldehyde, 1-phenyl, 1h-imidazole-2-carbaldehyde, 1-phenyl, 1-phenyl-2-formylimidazole, 1-phenylimidazole-2-carboxaldehyde
PubChem CID 6424701
IUPAC Name 1-phenylimidazole-2-carbaldehyde
SMILES C1=CC=C(C=C1)N2C=CN=C2C=O

Specifications

Packaging Amber Glass Bottle
Quantity 5 g
Formula Weight 172.19
Percent Purity 97%
Chemical Name or Material 1-phenyl-1H-imidazole-2-carbaldehyde
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.