Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Phenyl-1H-pyrazole-4-carboxylic acid, 99%

Catalog No. AAH50305MD
Change view
Click to view available options
Quantity:
250 mg
1 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAH50305MD 250 mg
AAH5030503 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAH50305MD Supplier Thermo Scientific Chemicals Supplier No. H50305MD
Only null left
Add to Cart

CAS: 1134-50-5 | C10H8N2O2 | 188.186 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1134-50-5
Molecular Formula C10H8N2O2
Molecular Weight (g/mol) 188.186
MDL Number MFCD00514518
InChI Key ZROILLPDIUNLSE-UHFFFAOYSA-N
Synonym 1-phenyl-1h-pyrazole-4-carboxylic acid, pyrazole-4-carboxylic acid, 1-phenyl, 1h-pyrazole-4-carboxylic acid, 1-phenyl, 1-phenylpyrazole-4-carboxylate, 1-phenyl-1h-pyrazol-4-carboxylic acid, pyrazole, 1, 1-phenyl-1h-pyrazole-4-carboxylicacid, akos pao-0321, acmc-1c9e6
PubChem CID 121026
IUPAC Name 1-phenylpyrazole-4-carboxylic acid
SMILES C1=CC=C(C=C1)N2C=C(C=N2)C(=O)O

Specifications

Quantity 250 mg
Formula Weight 188.19
Percent Purity 99%
Chemical Name or Material 1-Phenyl-1H-pyrazole-4-carboxylic acid
Hazard Category H315-H319-H335
Hazard Statement GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c
RTECSNumber UQ6436000
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.