Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC (±)-1-phenyl-2,3,4,5-tetrahydro-(1H)-3-benzazepine-7,8-diol hydrochloride | 62717-42-4 | MFCD00069248 | 99.5% | 291.77 | C16H18ClNO2 | 100MG
SDP

Catalog No. 5000238668
Encompass_Preferred

SKF 38393 hydrochloride is a selective dopamine D1 receptor agonist for research use that activates D1-mediated signaling, increases intracellular cAMP, and is used in cell signaling and in vivo pharmacology studies.

  • Selective D1 receptor agonist with reported IC50 of 110 nM.
  • High purity (99.49%).
  • CAS number 62717-42-4 for unambiguous identification.
  • Molecular formula C16H18ClNO2; molecular weight 291.77 Da.
  • Suitable for in vitro receptor activation and in vivo pharmacology models.
  • Storage: solid at 4°C; in solvent store at -80°C (6 months) or -20°C (1 month).
  • For research use only; not for human or veterinary use.

Catalog No. 50-002-38668 Supplier Medchemexpress LLC Supplier No. HY12520A100MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

SKF 38393 hydrochloride is a selective dopamine D1 receptor agonist for research use that activates D1-mediated signaling, increases intracellular cAMP, and is used in cell signaling and in vivo pharmacology studies.

  • Selective D1 receptor agonist with reported IC50 of 110 nM.
  • High purity (99.49%).
  • CAS number 62717-42-4 for unambiguous identification.
  • Molecular formula C16H18ClNO2; molecular weight 291.77 Da.
  • Suitable for in vitro receptor activation and in vivo pharmacology models.
  • Storage: solid at 4°C; in solvent store at -80°C (6 months) or -20°C (1 month).
  • For research use only; not for human or veterinary use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.