Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1-phenyl-2,3,4,5-tetrahydro-1H-3-benzazepine-7,8-diol hydrochloride | 62717-42-4 | MFCD00069248 | 99.5% | 291.77 g/mol | C16H18ClNO2 | 500MG
SDP

Catalog No. 5000238567
Encompass_Preferred

SKF 38393 hydrochloride is a selective agonist of the dopamine D1 receptor used as a research reagent to study D1-mediated signaling. It is supplied as a solid (also available as a 10 mM solution in DMSO) with analytical-grade purity and intended for research use only.

  • CAS number 62717-42-4.
  • Chemical formula C16H18ClNO2.
  • Molecular weight 291.77 g/mol.
  • D1 receptor IC50 ~110 nM.
  • Purity approximately 99.5%.
  • Available as solid quantities including 500 mg and larger sizes.
  • Provided for research use only; not for human or clinical use.

Catalog No. 50-002-38567 Supplier Medchemexpress LLC Supplier No. HY12520A500MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

SKF 38393 hydrochloride is a selective agonist of the dopamine D1 receptor used as a research reagent to study D1-mediated signaling. It is supplied as a solid (also available as a 10 mM solution in DMSO) with analytical-grade purity and intended for research use only.

  • CAS number 62717-42-4.
  • Chemical formula C16H18ClNO2.
  • Molecular weight 291.77 g/mol.
  • D1 receptor IC50 ~110 nM.
  • Purity approximately 99.5%.
  • Available as solid quantities including 500 mg and larger sizes.
  • Provided for research use only; not for human or clinical use.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.