Learn More
Medchemexpress LLC 1-phenyl-2,5,8,11,14-pentaoxahexadecan-16-ol | 57671-28-0 | MFCD06797154 | 98.6% | 328.40 g/mol | C17H28O6 | 1 G
SDP
Catalog No. 5000210385
Encompass_Preferred
BnO-PEG5-OH is a benzyl-terminated PEG5 primary alcohol used as a flexible linker in PROTAC synthesis. It provides a short polyethylene glycol spacer with a terminal benzyl-protected alcohol functionality that can be modified for conjugation to ligands and linkers.
  • Provides a PEG5 spacer for flexibility and improved solubility.
  • Benzyl-protected terminal alcohol enables selective deprotection and further functionalization.
  • High purity suitable for medicinal chemistry and research use.
  • Characterized by certificate of analysis, 1H NMR, and RP-HPLC documentation.
  • Available in small pack sizes for synthetic applications.
Catalog No. 50-002-10385 Supplier Medchemexpress LLC Supplier No. HYW0427141G
BnO-PEG5-OH is a benzyl-terminated PEG5 primary alcohol used as a flexible linker in PROTAC synthesis. It provides a short polyethylene glycol spacer with a terminal benzyl-protected alcohol functionality that can be modified for conjugation to ligands and linkers.
  • Provides a PEG5 spacer for flexibility and improved solubility.
  • Benzyl-protected terminal alcohol enables selective deprotection and further functionalization.
  • High purity suitable for medicinal chemistry and research use.
  • Characterized by certificate of analysis, 1H NMR, and RP-HPLC documentation.
  • Available in small pack sizes for synthetic applications.