Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Phenyl-2-propanol, 98%

Catalog No. p-3211417
Change view
Click to view available options
Quantity:
5 g
25 g
Packaging:
Glass bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC183060050 5 g Glass bottle
AC183060250 25 g Glass bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC183060050 Supplier Thermo Scientific Chemicals Supplier No. 183060050
Only null left
Add to Cart

CAS: 14898-87-4 | C9H12O | 136.19 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 14898-87-4
Molecular Formula C9H12O
Molecular Weight (g/mol) 136.19
MDL Number MFCD00004540
InChI Key WYTRYIUQUDTGSX-UHFFFAOYNA-N
Synonym 1-phenyl-2-propanol, benzyl methyl carbinol, 2-hydroxy-1-phenylpropane, benzylmethylcarbinol, 2-propanol, 1-phenyl, benzeneethanol, .alpha.-methyl, alpha-methyl-phenethyl alcohol, benzeneethanol, alpha-methyl, alpha-methylbenzeneethanol, .alpha.-methylphenethyl alcohol
PubChem CID 94185
IUPAC Name 1-phenylpropan-2-ol
SMILES CC(O)CC1=CC=CC=C1

Specifications

CAS Min % 98.5
CAS Max % 100.0
Color Colorless
Density 0.9900g/mL
Flash Point 85°C
Infrared Spectrum Authentic
Assay Percent Range 98.5% min. (GC)
Packaging Glass bottle
Refractive Index 1.5200 to 1.522
Linear Formula C6H5CH2CH(OH)CH3
Quantity 5 g
Beilstein 06,503
Specific Gravity 0.99
Formula Weight 136.19
Percent Purity 98%
Physical Form Liquid
Chemical Name or Material 1-Phenyl-2-propanol
Show More Show Less
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement:
Harmful if swallowed.
Precautionary Statement GHS P Statement:
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.