Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

1-Phenyl-2-propanol 98.0+%, TCI America™
SDP

Catalog No. P02155G Shop All Tokyo Chemical Industry (TCI) Products
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
P02155G 5 g
P021525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. P02155G Supplier TCI America Supplier No. P02155G
Add to Cart
Add to Cart

Chemical Identifiers

CAS 698-87-3
Molecular Formula C9H12O
Molecular Weight (g/mol) 136.19
MDL Number MFCD00004540
InChI Key WYTRYIUQUDTGSX-UHFFFAOYNA-N
Synonym Benzylmethylcarbinol
PubChem CID 94185
IUPAC Name 1-phenylpropan-2-ol
SMILES CC(O)CC1=CC=CC=C1

Specifications

Color Colorless
Boiling Point 213°C
Quantity 5 g
Formula Weight 136.19
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material 1-Phenyl-2-propanol
EINECSNumber (3)-1034
TSCA Yes
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.