Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1-Phenyl-2-trimethylsilylacetylene

Catalog No. 33892350G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 g
50 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
33-892-350G 50 g
33-892-310G 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 33-892-350G Supplier Sigma Aldrich Supplier No. 33892350G
  • NEW LOWER PRICE
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • 1-phenyl-2-trimethylsilylacetylene is an sime3-substituted alkyne
  • It undergoes coupling with propargylic and allylic alcohols
  • Polymerization of 1-phenyl-2-trimethylsilylacetylene by various transition metal catalysts has been reported.1-phenyl-2-trimethylsilylacetylene was used in the preparation of donor-stabilized pt-++2-alkyne complexes

Specifications

Density 0.886 g/mL (at 25°C (literature))
Boiling Point 87°C to 88°C (9 mmHg)
Molecular Formula C11H14Si
Refractive Index n20/D 1.528 (literature)
Linear Formula C6H5C///CSi(CH3)3
MDL Number MFCD00054860
Quantity 50 g
Synonym 1-(Trimethylsilyl)-2-phenylacetylene; Phenylethynyl-trimethylsilane; Trimethyl(phenylethynyl)silane
Molecular Weight (g/mol) 174.31
Percent Purity 99%
Biological Activity Respiratory System
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.