Learn More
1-Phenyl-3-pyrazolidinone, 97%
Description
Phenidone inhibits both the LO (lipoxygenase) and Cox (cyclooxygenase) pathways, the synthesis of Fos-related antigen protein, and is described as an anti-inflammatory and anti-oxidant compound. Phenidone has been shown to block neurotoxicity induced by kainate, and weakens oxidative stress induced by lipopolysaccharide.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Phenidone inhibits both the LO (lipoxygenase) and Cox (cyclooxygenase) pathways, the synthesis of Fos-related antigen protein, and is described as an anti-inflammatory and anti-oxidant compound. Phenidone has been shown to block neurotoxicity induced by kainate, and weakens oxidative stress induced by lipopolysaccharide.
Solubility
Soluble in water (1:10 hot), methanol (0.1 mg/mL), DMSO (75 mg/mL), and ethanol.
Notes
Store in cool place. Keep away from oxidizing agents.
Specifications
Specifications
| Melting Point | 120°C to 123°C |
| Quantity | 1000 g |
| UN Number | UN2811 |
| Beilstein | 131856 |
| Sensitivity | Light sensitive |
| Merck Index | 14,7309 |
| Solubility Information | Soluble in water (1:10 hot),methanol (0.1mg/mL),DMSO (75mg/mL),and ethanol. |
| Formula Weight | 162.19 |
| Percent Purity | 97% |
| Chemical Name or Material | 1-Phenyl-3-pyrazolidinone |
Safety and Handling
| Hazard Category | H301 |
| Hazard Statement | GHS H Statement H302 Harmful if swallowed. |
| Precautionary Statement | P264b-P270-P301+P310-P330-P501c |
| DOTInformation | Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S. |
| EINECSNumber | 202-155-1 |
| RTECSNumber | UQ8750000 |
| TSCA | Yes |
| Recommended Storage | Ambient temperatures |
RUO – Research Use Only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.