Learn More
Selleck Chemical LLC 1-Phenylbiguanide S2578-25mg
SDP
Catalog No. 503112404
Encompass_Preferred
1-Phenylbiguanide (Phenylbiguanide PBG N-Phenylbiguanide) is a 5-hydroxytryptamine3 (5-HT3) receptor agonist with EC50 of 3 0 M
Catalog No. 50-311-2404 Supplier Selleck Chemical LLC Supplier No. S257825MG
1-Phenylbiguanide (Phenylbiguanide PBG N-Phenylbiguanide) is a 5-hydroxytryptamine3 (5-HT3) receptor agonist with EC50 of 3 0 M