Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich 1-Phenylcyclopentanamine hydrochloride

Catalog No. OTV0006121G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
25 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
OTV0006121G 25 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. OTV0006121G Supplier Sigma Aldrich Supplier No. OTV0006121G
Only null left
Add to Cart
Add to Cart

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

  • Dimethylphosphoramidous dichloride (dimethylaminodichlorophosphine) is a phosphorus chloride derivative. it exists in two stable isomers, a gauche-conformer with c1 symmetry and anti-conformer with cs symmetry. its synthesis has been reported. molecular conformation has been investigated through correlation between the 35cl nqr frequencies and charge on the chlorine atoms. its vibrational spectra has been studied. its reaction with so2 has been investigated. it participates in the synthesis of [(dimethylamino)phosphoryl] bis(difluoromethylene)phosphonate.dimethylphosphoramidous dichloride (n,n-dimethylamidodichlorophosphite) may be used in the synthesis of the following:
  • N,n-dimethylamidotetrachlorophosphorane
  • O-propylchloroformimino-n,n-dimethylamidochlorophosphate
  • 1,3-bisarylsulphonyl-1,3,2,4-diazadiphosphetidines

Specifications

CAS 5296-90-2
Molecular Formula C11H16ClN
Linear Formula C11H16ClN
MDL Number MFCD12914020
Quantity 25 mL
Molecular Weight (g/mol) 197.7
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.