Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1-Phenylethane-1,2-diol | 93-56-1 | MFCD00003546 | 99.7% | 138.17 g/mol | C8H10O2 | 10 G
SDP

Catalog No. 5000300756
Encompass_Preferred

1-Phenylethane-1,2-diol (styrene glycol) is a benzyl diol used as an analytical standard and research reagent. It is a major metabolite of styrene and is commonly employed in analytical chemistry, metabolic studies, and method development. The product is supplied as a solid with documented batch purity and supporting certificate of analysis for quality assurance.

  • Provides high purity for analytical applications (HPLC 99.74%).
  • Solid form, off-white to yellow appearance.
  • Molecular formula C8H10O2; molecular weight 138.17 g/mol.
  • Suitable as an analytical standard and for metabolic study workflows.
  • Batch certificate of analysis available for traceability.
  • Supplied in small research-friendly package sizes, including 10 g.

Catalog No. 50-003-00756 Supplier Medchemexpress LLC Supplier No. HYW01578810G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

1-Phenylethane-1,2-diol (styrene glycol) is a benzyl diol used as an analytical standard and research reagent. It is a major metabolite of styrene and is commonly employed in analytical chemistry, metabolic studies, and method development. The product is supplied as a solid with documented batch purity and supporting certificate of analysis for quality assurance.

  • Provides high purity for analytical applications (HPLC 99.74%).
  • Solid form, off-white to yellow appearance.
  • Molecular formula C8H10O2; molecular weight 138.17 g/mol.
  • Suitable as an analytical standard and for metabolic study workflows.
  • Batch certificate of analysis available for traceability.
  • Supplied in small research-friendly package sizes, including 10 g.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.