Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Sigma Aldrich ™-1-Phenylethanol

Catalog No. 6858285G Shop All MilliporeSigma Sigma Organic Chemistry Products
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
68-582-85G 5 g
68-582-825G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Warning
Catalog No. 68-582-85G Supplier Sigma Aldrich Supplier No. 6858285G
May include imposed supplier surcharges.
Add to Cart
Add to Cart
Warning

MilliporeSigma Sigma Organics products encompass a wide range of quality reagents, solvents, catalysts, and building blocks for organic synthesis. From benchtop discovery to process development and scale-up, Sigma Organics solutions are built to meet the needs of synthetic chemists.

(R)-1-Phenylethanol can be prepared from ethylbenzene via enantioselective hydroxylation of ethylbenzene catalyzed by peroxygenase enzyme.

Specifications

CAS 1517-69-7
Density 0.9986 g/mL at 25°C
Molecular Formula C8H10O
Refractive Index n20/D 1.528
Linear Formula C6H5CH(OH)CH3
MDL Number MFCD00064263
Quantity 5 g
Molecular Weight (g/mol) 122.16 g/mol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.