Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1-phenylethyl propionate | 120-45-6 | MFCD00009305 | 178.23 g·mol⁻1 | C11H14O2 | 1 KG
SDP

Supplier:  Medchemexpress LLC HYW0165981KG

Encompass_Preferred

1-Phenylethyl propionate (CAS 120-45-6) is an aromatic ester reagent (C11H14O2, MW 178.23 g·mol⁻1) commonly used in organic synthesis and biochemical assay development. It is supplied as a colorless to light yellow liquid; consult the SDS for handling, storage, and safety information.

  • Used as an ester reagent in organic synthesis and assay development.
  • Typical physical form: colorless to light yellow liquid.
  • Molecular formula C11H14O2 and molecular weight 178.23 g·mol⁻1.
  • Available in multiple pack sizes to support small- to large-scale work.
  • Storage and stability guidance provided in the SDS for long-term retention.

Catalog No. 50-002-99377


May include imposed supplier surcharges.
Add to Cart

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.