Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1-phenylethyl propionate | 120-45-6 | MFCD00009305 | 178.23 g/mol | C11H14O2 | 25 G
SDP

Catalog No. 5000303486
Encompass_Preferred

1-Phenylethyl propionate is an aromatic ester used as a biochemical reagent and intermediate in organic synthesis. Supplied for research use only, it is commonly used in assay development, synthetic transformations, and fragrance component studies. Consult the safety data sheet for handling, storage, and disposal instructions.

  • Aromatic ester with formula C11H14O2.
  • Molecular weight 178.23 g/mol.
  • Typically a clear, colorless liquid.
  • Soluble in common organic solvents.
  • Available in multiple lab-scale pack sizes for synthesis and assays.
  • Research use only; follow SDS and safety procedures.

Catalog No. 50-003-03486 Supplier Medchemexpress LLC Supplier No. 25GHYW016598
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

1-Phenylethyl propionate is an aromatic ester used as a biochemical reagent and intermediate in organic synthesis. Supplied for research use only, it is commonly used in assay development, synthetic transformations, and fragrance component studies. Consult the safety data sheet for handling, storage, and disposal instructions.

  • Aromatic ester with formula C11H14O2.
  • Molecular weight 178.23 g/mol.
  • Typically a clear, colorless liquid.
  • Soluble in common organic solvents.
  • Available in multiple lab-scale pack sizes for synthesis and assays.
  • Research use only; follow SDS and safety procedures.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.