Learn More
Medchemexpress LLC 1-phenylethyl propionate | 120-45-6 | MFCD00009305 | >98.0% | 178.23 g/mol | C11H14O2 | 100 G
SDP
Catalog No. 5000303400
Encompass_Preferred
1-Phenylethyl propionate (CAS 120-45-6) is an organic ester used as a biochemical reagent and as an intermediate in fragrance and flavor applications. Supplied for research use, it is typically a clear liquid with molecular formula C11H14O2 and molecular weight 178.23 g/mol.
  • High purity (>98%) suitable for analytical and research use.
  • Useful as a biochemical reagent and fragrance/flavor intermediate.
  • Liquid at room temperature; density approximately 1.0 g/cm3.
  • Available in multiple pack sizes to support lab workflows.
  • CAS identifier provided for unambiguous substance matching.
Catalog No. 50-003-03400 Supplier Medchemexpress LLC Supplier No. HYW016598100G
1-Phenylethyl propionate (CAS 120-45-6) is an organic ester used as a biochemical reagent and as an intermediate in fragrance and flavor applications. Supplied for research use, it is typically a clear liquid with molecular formula C11H14O2 and molecular weight 178.23 g/mol.
  • High purity (>98%) suitable for analytical and research use.
  • Useful as a biochemical reagent and fragrance/flavor intermediate.
  • Liquid at room temperature; density approximately 1.0 g/cm3.
  • Available in multiple pack sizes to support lab workflows.
  • CAS identifier provided for unambiguous substance matching.