Learn More
Medchemexpress LLC 1-phenylethyl propionate | 120-45-6 | MFCD00009305 | >98.0% | 178.23 g/mol | C11H14O2 | 50 G
SDP
Catalog No. 5000301124
Encompass_Preferred
1-Phenylethyl propionate is a biochemical reagent (CAS 120-45-6) with molecular formula C11H14O2 and molecular weight 178.23 g/mol. It is supplied as a liquid with a reported density of about 1.0 ± 0.1 g/cm3 and is offered at high purity for research applications.
  • Has CAS number 120-45-6.
  • Molecular formula C11H14O2 and molecular weight 178.23 g/mol.
  • Liquid physical state with density 1.0 ± 0.1 g/cm3.
  • High purity (">98%") suitable for research use.
  • Provided in laboratory pack sizes for analytical and preparative workflows.
Catalog No. 50-003-01124 Supplier Medchemexpress LLC Supplier No. HYW01659850G
1-Phenylethyl propionate is a biochemical reagent (CAS 120-45-6) with molecular formula C11H14O2 and molecular weight 178.23 g/mol. It is supplied as a liquid with a reported density of about 1.0 ± 0.1 g/cm3 and is offered at high purity for research applications.
  • Has CAS number 120-45-6.
  • Molecular formula C11H14O2 and molecular weight 178.23 g/mol.
  • Liquid physical state with density 1.0 ± 0.1 g/cm3.
  • High purity (">98%") suitable for research use.
  • Provided in laboratory pack sizes for analytical and preparative workflows.