Learn More
Medchemexpress LLC 1-phenylpropane-1,2-dione | 579-07-7 | MFCD00008755 | 99.5% | 148.16 g·mol⁻¹ | C9H8O2 | 10 G
SDP
Catalog No. 5000301083
Encompass_Preferred
1-Phenylpropane-1,2-dione (CAS 579-07-7) is an aromatic alpha-diketone used in research chemistry and natural product studies. It is reported as a biosynthetic precursor to ephedrine alkaloids and is supplied as a high-purity reagent for laboratory and analytical applications. Intended for research use only, the material is provided with documentation to support safe handling and use.
  • High purity suitable for analytical and synthetic applications.
  • Alpha-diketone structure useful as a biosynthetic intermediate.
  • Available in multiple quantities and formats for flexible use.
  • Provided with purity documentation and a safety data sheet.
  • Intended for research use only; not for clinical or human use.
Catalog No. 50-003-01083 Supplier Medchemexpress LLC Supplier No. HYW01875810G
1-Phenylpropane-1,2-dione (CAS 579-07-7) is an aromatic alpha-diketone used in research chemistry and natural product studies. It is reported as a biosynthetic precursor to ephedrine alkaloids and is supplied as a high-purity reagent for laboratory and analytical applications. Intended for research use only, the material is provided with documentation to support safe handling and use.
  • High purity suitable for analytical and synthetic applications.
  • Alpha-diketone structure useful as a biosynthetic intermediate.
  • Available in multiple quantities and formats for flexible use.
  • Provided with purity documentation and a safety data sheet.
  • Intended for research use only; not for clinical or human use.