Learn More
Medchemexpress LLC 1-phenylpropane-1,2-dione | 579-07-7 | MFCD00008755 | 99.8% | 148.16 g/mol | C9H8O2 | 25 G
SDP
Catalog No. 5000302083
Encompass_Preferred
1-Phenylpropane-1,2-dione (CAS 579-07-7) is an alpha-diketone reported as a biosynthetic precursor to ephedrine alkaloids. Supplied for research use, it appears as a light yellow to yellow liquid with a molecular weight of 148.16 g/mol and is provided in solutions and bulk quantities. Handling recommendations include cold, dark storage and inert atmosphere for long-term stability.
  • High purity (~99.8%) suitable for analytical and synthetic work.
  • Available in multiple formats, including concentrated DMSO solution.
  • Light yellow to yellow liquid, density approximately 1.101 g/cm3.
  • Recommended storage at 4°C, protect from light, store under inert gas for stability.
  • Relevant as a biosynthetic intermediate for alkaloid research applications.
Catalog No. 50-003-02083 Supplier Medchemexpress LLC Supplier No. 25GHYW018758
1-Phenylpropane-1,2-dione (CAS 579-07-7) is an alpha-diketone reported as a biosynthetic precursor to ephedrine alkaloids. Supplied for research use, it appears as a light yellow to yellow liquid with a molecular weight of 148.16 g/mol and is provided in solutions and bulk quantities. Handling recommendations include cold, dark storage and inert atmosphere for long-term stability.
  • High purity (~99.8%) suitable for analytical and synthetic work.
  • Available in multiple formats, including concentrated DMSO solution.
  • Light yellow to yellow liquid, density approximately 1.101 g/cm3.
  • Recommended storage at 4°C, protect from light, store under inert gas for stability.
  • Relevant as a biosynthetic intermediate for alkaloid research applications.