Learn More
Medchemexpress LLC 1-phenylpropane-1,2-dione | 579-07-7 | MFCD00008755 | 99.8% | 148.16 g/mol | C9H8O2 | 5 G
SDP
Catalog No. 5000301084
Encompass_Preferred
1-Phenylpropane-1,2-dione is an alpha-diketone (C9H8O2; MW 148.16 g/mol) isolated from young Ephedra sinica and reported as a biosynthetic precursor to ephedrine alkaloids. It is provided as a research reagent for analytical and synthetic applications.
  • High purity (≈99.8%) for reliable experimental results.
  • Light yellow liquid with density about 1.101 g/cm3.
  • Highly soluble in DMSO (≈200 mg/mL), suitable for in vitro preparations.
  • Storage: keep at 4°C, protect from light, and store under inert gas; solutions stable at -80°C for up to 6 months.
  • Available in multiple laboratory pack sizes to support small- and bulk-scale work.
Catalog No. 50-003-01084 Supplier Medchemexpress LLC Supplier No. HYW0187585G
1-Phenylpropane-1,2-dione is an alpha-diketone (C9H8O2; MW 148.16 g/mol) isolated from young Ephedra sinica and reported as a biosynthetic precursor to ephedrine alkaloids. It is provided as a research reagent for analytical and synthetic applications.
  • High purity (≈99.8%) for reliable experimental results.
  • Light yellow liquid with density about 1.101 g/cm3.
  • Highly soluble in DMSO (≈200 mg/mL), suitable for in vitro preparations.
  • Storage: keep at 4°C, protect from light, and store under inert gas; solutions stable at -80°C for up to 6 months.
  • Available in multiple laboratory pack sizes to support small- and bulk-scale work.
missing translation for 'productTitle'
missing translation for 'pleaseSelectIssue'
missing translation for 'byClickingSubmit' missing translation for 'privacyPolicy'.