Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1-phenylpyrrole | 635-90-5 | MFCD00005343 | 99.9% | 143.19 g/mol | C10H9N | 1G
SDP

Catalog No. 5000239501
Encompass_Preferred

1-Phenylpyrrole is a small aromatic heterocycle (C10H9N) used as a fluorescent probe and synthetic building block in research. It exhibits dual fluorescence-a single emission band in nonpolar solvents and a red-shifted secondary band in polar solvents. Supplied as a high-purity solid, it is soluble in DMSO and has defined storage recommendations for powder and solutions.

  • Dual fluorescent behavior in polar and nonpolar solvents.
  • High purity (99.94%).
  • Molecular weight 143.19 g/mol, formula C10H9N.
  • Readily soluble in DMSO (100 mg/mL) and formulatable for in vivo dosing.
  • Supplied as a solid in multiple pack sizes including 1 g.
  • Storage recommendations for powder and solutions provided.

Catalog No. 50-002-39501 Supplier Medchemexpress LLC Supplier No. HY1248911G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

1-Phenylpyrrole is a small aromatic heterocycle (C10H9N) used as a fluorescent probe and synthetic building block in research. It exhibits dual fluorescence-a single emission band in nonpolar solvents and a red-shifted secondary band in polar solvents. Supplied as a high-purity solid, it is soluble in DMSO and has defined storage recommendations for powder and solutions.

  • Dual fluorescent behavior in polar and nonpolar solvents.
  • High purity (99.94%).
  • Molecular weight 143.19 g/mol, formula C10H9N.
  • Readily soluble in DMSO (100 mg/mL) and formulatable for in vivo dosing.
  • Supplied as a solid in multiple pack sizes including 1 g.
  • Storage recommendations for powder and solutions provided.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.