Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1-phenylpyrrole | 635-90-5 | MFCD00005343 | 99.9% | 143.19 g/mol | C10H9N | 5G
SDP

Catalog No. 5000236529
Encompass_Preferred

1-Phenylpyrrole is a small organic compound that exhibits polarity-dependent dual fluorescence, showing a single emission band in nonpolar solvents and a red-shifted band in polar solvents. It is supplied as a solid and as a concentrated DMSO solution for use in photophysical and fluorescence studies.

  • Exhibits dual fluorescence dependent on solvent polarity.
  • High purity (99.94%) suitable for research applications.
  • Molecular formula C10H9N; molecular weight 143.19 g/mol.
  • Available as solid and 10 mM solution in DMSO; DMSO solubility 100 mg/mL.
  • Physical appearance: light yellow to light brown solid.
  • Storage: powder at -20°C (up to 3 years) or 4°C (up to 2 years); stock solutions at -80°C (6 months).
  • Intended for fluorescence and solvent-dependent photophysical investigations.

Catalog No. 50-002-36529 Supplier Medchemexpress LLC Supplier No. HY1248915G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

1-Phenylpyrrole is a small organic compound that exhibits polarity-dependent dual fluorescence, showing a single emission band in nonpolar solvents and a red-shifted band in polar solvents. It is supplied as a solid and as a concentrated DMSO solution for use in photophysical and fluorescence studies.

  • Exhibits dual fluorescence dependent on solvent polarity.
  • High purity (99.94%) suitable for research applications.
  • Molecular formula C10H9N; molecular weight 143.19 g/mol.
  • Available as solid and 10 mM solution in DMSO; DMSO solubility 100 mg/mL.
  • Physical appearance: light yellow to light brown solid.
  • Storage: powder at -20°C (up to 3 years) or 4°C (up to 2 years); stock solutions at -80°C (6 months).
  • Intended for fluorescence and solvent-dependent photophysical investigations.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.