Learn More
Medchemexpress LLC 1-phenylpyrrole (N-phenylpyrrole) | 635-90-5 | MFCD00005343 | 99.9% | 143.18 g/mol | C10H9N | 10MM 1ML
SDP
Catalog No. 5000235627
Encompass_Preferred
1-Phenylpyrrole (N-Phenylpyrrole) is an aromatic heterocycle that exhibits solvent-dependent dual fluorescence, showing a single emission band in nonpolar solvents and a red-shifted band in polar solvents. It is commonly used as a fluorescent probe and in spectroscopic studies. The compound is supplied both as a 10 mM solution in DMSO and as solid material, with reported high purity.
Catalog No. 50-002-35627 Supplier Medchemexpress LLC Supplier No. HY12489110MM 1ML
1-Phenylpyrrole (N-Phenylpyrrole) is an aromatic heterocycle that exhibits solvent-dependent dual fluorescence, showing a single emission band in nonpolar solvents and a red-shifted band in polar solvents. It is commonly used as a fluorescent probe and in spectroscopic studies. The compound is supplied both as a 10 mM solution in DMSO and as solid material, with reported high purity.