Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1-phenylpyrrole (N-phenylpyrrole) | 635-90-5 | MFCD00005343 | 99.9% | 143.18 g/mol | C10H9N | 25G
SDP

Catalog No. 5000239502
Encompass_Preferred

1-Phenylpyrrole is an aromatic heterocycle that exhibits solvent-dependent dual fluorescence, showing a single emission band in nonpolar solvents and a red-shifted band in polar solvents. It is supplied as a solid or as solutions (for example, 10 mM in DMSO), is reported with high purity, and is used in photophysical and solvent polarity studies.

  • Exhibits solvent-dependent dual fluorescence suitable for photophysical studies.
  • High reported purity (approximately 99.9%).
  • Solid appearance: light yellow to light brown powder.
  • Available as solutions and multiple solid package sizes, including 25 g.
  • Molecular formula C10H9N and molecular weight ~143.18 g/mol.
  • Recommended storage: powder at -20°C (long term) or 4°C (short term); in solvent at -80°C for extended periods.
  • Shipped at room temperature in some regions; conditions may vary by location.

Catalog No. 50-002-39502 Supplier Medchemexpress LLC Supplier No. HY12489125G
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

1-Phenylpyrrole is an aromatic heterocycle that exhibits solvent-dependent dual fluorescence, showing a single emission band in nonpolar solvents and a red-shifted band in polar solvents. It is supplied as a solid or as solutions (for example, 10 mM in DMSO), is reported with high purity, and is used in photophysical and solvent polarity studies.

  • Exhibits solvent-dependent dual fluorescence suitable for photophysical studies.
  • High reported purity (approximately 99.9%).
  • Solid appearance: light yellow to light brown powder.
  • Available as solutions and multiple solid package sizes, including 25 g.
  • Molecular formula C10H9N and molecular weight ~143.18 g/mol.
  • Recommended storage: powder at -20°C (long term) or 4°C (short term); in solvent at -80°C for extended periods.
  • Shipped at room temperature in some regions; conditions may vary by location.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.