Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

[1-(Phenylsulfonyl)-1H-indol-3-yl]methanol, ≥97%, Thermo Scientific™

Catalog No. CC03109CB
Change view
Click to view available options
Quantity:
250 mg
1 g
Packaging:
Amber Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
CC03109CB 250 mg Amber Glass Bottle
CC03109DA 1 g Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. CC03109CB Supplier Thermo Scientific Chemicals Supplier No. CC03109CB
Add to Cart

Chemical Identifiers

CAS 89241-33-8
Molecular Formula C15H13NO3S
Molecular Weight (g/mol) 287.333
MDL Number MFCD02682024
InChI Key ZMLXSFMIPYDHIN-UHFFFAOYSA-N
Synonym 1-phenylsulfonyl-1h-indol-3-yl methanol, 1-benzenesulfonyl indol-3-yl methanol, 1h-indole-3-methanol,1-phenylsulfonyl, 1-benzenesulfonyl-1h-indol-3-yl methanol, acmc-20ljqe, 1-benzenesulfonyl-1h-indol-3-ylmethanol, 3-hydroxymethyl-n-phenylsulfonyl indole, 1-phenylsulfonyl-1h-indole-3-methanol
PubChem CID 2776213
IUPAC Name [1-(benzenesulfonyl)indol-3-yl]methanol
SMILES C1=CC=C(C=C1)S(=O)(=O)N2C=C(C3=CC=CC=C32)CO

Specifications

CAS Min % 96.5
CAS Max % 100.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 250 mg
Formula Weight 287.34
Percent Purity 97%
Chemical Name or Material [1-(Phenylsulfonyl)-1H-indol-3-yl]methanol
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.