Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC 1-Piperazineacetamide, 4-[6-[(6-acetyl-8-cyclopentyl-7,8-dihydro-5-methyl-7-oxopyrido[2,3-d]pyrimidin-2-yl)amino]-3-pyridinyl]-N-[6-[[3-[[[6-(dimethylamino)-3-pyridazinyl]amino]carbonyl]-6-(methylamino)-2-pyridinyl]amino]spiro[3.3]hept-2-yl]- | 3055472-38-0 | 98.1% | 884.04 | 25 MG
SDP

Catalog No. 5000394292
Encompass_Preferred

CPD-39 is a potent and orally active CCND1 and CDK4 heterobifunctional PROTAC degrader that shows anti-proliferation. It is intended for research use only and is not sold to patients.

  • Target: CDK4, CCND1
  • Appearance: Solid, colorless to light yellow
  • Shipping: Room temperature in continental US; may vary elsewhere
  • Storage: Powder: -20°C for 3 years, 4°C for 2 years
  • Storage: In solvent: -80°C for 6 months, -20°C for 1 month

Catalog No. 50-003-94292 Supplier Medchemexpress LLC Supplier No. 25MGHY168664
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

CPD-39 is a potent and orally active CCND1 and CDK4 heterobifunctional PROTAC degrader that shows anti-proliferation. It is intended for research use only and is not sold to patients.

  • Target: CDK4, CCND1
  • Appearance: Solid, colorless to light yellow
  • Shipping: Room temperature in continental US; may vary elsewhere
  • Storage: Powder: -20°C for 3 years, 4°C for 2 years
  • Storage: In solvent: -80°C for 6 months, -20°C for 1 month

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.