Learn More
Medchemexpress LLC 1-Piperazinecarboxylic acid, 2-(cyanomethyl)-4-[5,6,7,8-tetrahydro-2-[[(2S,4R)-4-[2-[2-[2-(2-hydroxyethoxy)ethoxy]ethoxy]ethoxy]-1-methyl-2-pyrrolidinyl]methoxy]-7-(1-naphthalenyl)pyrido[3,4-d]pyrimidin-4-yl]- | 2378261-85-7 | 99.8% | 25 MG
SDP
Catalog No. 5000341421
Encompass_Preferred
This K-Ras ligand-Linker Conjugate 5 is a target protein ligand-linker conjugate with a high purity of 99.79% (LCMS). It appears as a light yellow to yellow solid and has a molecular formula of C42H59N7O8 and a molecular weight of 789.96. It is intended for research use only.
  • High purity (99.79%)
  • Light yellow to yellow solid appearance
  • Molecular weight of 789.96
  • Suitable for research applications
Catalog No. 50-003-41421 Supplier Medchemexpress LLC Supplier No. 25MGHY130823
This K-Ras ligand-Linker Conjugate 5 is a target protein ligand-linker conjugate with a high purity of 99.79% (LCMS). It appears as a light yellow to yellow solid and has a molecular formula of C42H59N7O8 and a molecular weight of 789.96. It is intended for research use only.
  • High purity (99.79%)
  • Light yellow to yellow solid appearance
  • Molecular weight of 789.96
  • Suitable for research applications