Learn More
Medchemexpress LLC 1-piperazinedodecanamide, phthalazinyl-fluorobenzoyl isoindole piperidinyl amide | 2523016-96-6 | 99.9% | C47H53FN8O8 | 10MG
SDP
Catalog No. 5000261068
Encompass_Preferred
SK-575 is a proteolysis-targeting chimera (PROTAC) that selectively induces degradation of PARP1. It is supplied as a research-grade small molecule for biochemical and cellular studies of PARP1 biology and targeted protein degradation.
  • Selective PARP1 degradation via PROTAC mechanism.
  • Potent activity with low-nanomolar efficacy.
  • High chemical purity (~99.9%) suitable for biological assays.
  • Available in research-friendly quantities for assay development.
  • Datasheet and certificate of analysis available for batch verification.
Catalog No. 50-002-61068 Supplier Medchemexpress LLC Supplier No. HY13915610MG
SK-575 is a proteolysis-targeting chimera (PROTAC) that selectively induces degradation of PARP1. It is supplied as a research-grade small molecule for biochemical and cellular studies of PARP1 biology and targeted protein degradation.
  • Selective PARP1 degradation via PROTAC mechanism.
  • Potent activity with low-nanomolar efficacy.
  • High chemical purity (~99.9%) suitable for biological assays.
  • Available in research-friendly quantities for assay development.
  • Datasheet and certificate of analysis available for batch verification.