Learn More
Medchemexpress LLC 1-Piperazineethanol, 4-[(3E)-3-(2-chloro-9H-thioxanthen-9-ylidene)propyl]- | 53772-84-2 | 99.4% | C22H25ClN2OS | 25 MG
SDP
Catalog No. 5000359052
Encompass_Preferred
Trans-Clopenthixol is an off-white to light yellow solid. It is an antibiotic that does not have a neuroleptic effect and is intended for research use only, as it has not been fully validated for medical applications.
  • Inhibits Pseudomonas aeruginosa and Plasmodium falciparum in vitro.
  • Product complies with given specifications.
Catalog No. 50-003-59052 Supplier Medchemexpress LLC Supplier No. 25MGHYA0163D
Trans-Clopenthixol is an off-white to light yellow solid. It is an antibiotic that does not have a neuroleptic effect and is intended for research use only, as it has not been fully validated for medical applications.
  • Inhibits Pseudomonas aeruginosa and Plasmodium falciparum in vitro.
  • Product complies with given specifications.