Learn More
Medchemexpress LLC 1-Piperazinepropanesulfonic acid, β-hydroxy-4-(2-hydroxyethyl)- | 68399-78-0 | 99.9% | 10 G
SDP
Catalog No. 5000410066
Encompass_Preferred
HEPPSO is a zwitterionic buffer. It has a working pH range of 7.1-8.5. HEPPSO demonstrates a relatively high capacity to bind copper(II) and has a pKa of 7.84 at a 2.0 mM buffer concentration.
  • Zwitterionic buffer
  • Working pH range of 7.1-8.5
  • High capacity to bind copper(II)
  • pKa of 7.84 at 2.0 mM buffer concentration
Catalog No. 50-004-10066 Supplier Medchemexpress LLC Supplier No. HYD087410G
HEPPSO is a zwitterionic buffer. It has a working pH range of 7.1-8.5. HEPPSO demonstrates a relatively high capacity to bind copper(II) and has a pKa of 7.84 at a 2.0 mM buffer concentration.
  • Zwitterionic buffer
  • Working pH range of 7.1-8.5
  • High capacity to bind copper(II)
  • pKa of 7.84 at 2.0 mM buffer concentration