Learn More
Medchemexpress LLC 1-Piperazinepropanesulfonic acid, β-hydroxy-4-(2-hydroxyethyl)- | 68399-78-0 | 99.9% | 50 G
SDP
Catalog No. 5000410622
Encompass_Preferred
HEPPSO is a zwitterionic buffer with a working pH range of 7.1-8.5. It has a pKa of 7.84 at 2.0 mM buffer concentration and exhibits a relatively high ability to bind copper(II). It is for research use only.
  • Zwitterionic buffer
  • Working pH range: 7.1-8.5
  • pKa of 7.84 at 2.0 mM buffer concentration
  • Relatively high ability to bind copper(II)
Catalog No. 50-004-10622 Supplier Medchemexpress LLC Supplier No. HYD087450G
HEPPSO is a zwitterionic buffer with a working pH range of 7.1-8.5. It has a pKa of 7.84 at 2.0 mM buffer concentration and exhibits a relatively high ability to bind copper(II). It is for research use only.
  • Zwitterionic buffer
  • Working pH range: 7.1-8.5
  • pKa of 7.84 at 2.0 mM buffer concentration
  • Relatively high ability to bind copper(II)